missing translation for 'onlineSavingsMsg'
Learn More

Forsterite, Thermo Scientific™

Product Code. 11345347
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345347 5 g 5g
11355347 1 g 1g
11335347 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11345347 Supplier Thermo Scientific Alfa Aesar Supplier No. 042639.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15118-03-3
Molecular Formula H4Mg2O4Si
Molecular Weight (g/mol) 144.72
MDL Number MFCD00283278
InChI Key ICUHFTWXYNJXLX-UHFFFAOYSA-N
IUPAC Name silicic acid dimagnesium
SMILES [Mg].[Mg].O[Si](O)(O)O

Specifications

Color Green
Physical Form Grains
Quantity 5 g
Solubility Information Soluble in hot HCl. Insoluble in water.
Formula Weight 140.71
Odor Odorless
Refractive Index 1.63
Appearance Light green chunks
Density 3.21g/cm3
Chemical Name or Material Forsterite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.