accessibility menu, dialog, popup

UserName

MgMgOHSiOHOHHO

CAS RN 15118-03-3

IUPAC Name: silicic acid dimagnesium
Synonyms: silicic acid dimagnesium
Molecular Weight (g/mol): 144.72
Molecular Formula: H4Mg2O4Si
InChi Key: ICUHFTWXYNJXLX-UHFFFAOYSA-N
SMILES: [Mg].[Mg].O[Si](O)(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results