Learn More
Forskolin, 98+%, Thermo Scientific™
Forskolin, CAS # 66575-29-9, is a diterpenoid compound isolated from Indian Coleus that acts as a strong activator of the adenylate cyclase system.
£123.65 - £495.65
Chemical Identifiers
| CAS | 66575-29-9 |
|---|---|
| Molecular Formula | C22H34O7 |
| Molecular Weight (g/mol) | 410.507 |
| MDL Number | MFCD00082317 |
| InChI Key | OHCQJHSOBUTRHG-KGGHGJDLSA-N |
| Synonym | forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish |
| PubChem CID | 47936 |
| ChEBI | CHEBI:42471 |
| IUPAC Name | [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate |
| SMILES | CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15435559
|
Thermo Scientific Alfa Aesar
J63292.MA |
10mg |
£137.00 £123.65 / 10mg Save £13.35 9% Off |
In Stock
Log in to see stock.
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15445559
|
Thermo Scientific Alfa Aesar
J63292.MF |
50mg |
£550.00 £495.65 / 50mg Save £54.35 9% Off |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Forskolin is a diterpenoid and cyclic ketone, found naturally as a plant metabolite
• It is a protein kinase A agonist and a cyclic ketone • Forskolin is an antihypertensive and muscle relaxant; it can also promote the release of pituitary hormones and prevent platelet aggregation
Application
• Forskolin assists in neuron differentiation in vitro
• Useful for studies of cell metabolism and development; there is some evidence of its usefulness in the study of glaucoma
• It is intended for cell culture and biochemistry applications
Chemical Identifiers
| 66575-29-9 | |
| 410.507 | |
| OHCQJHSOBUTRHG-KGGHGJDLSA-N | |
| 47936 | |
| [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate |
| C22H34O7 | |
| MFCD00082317 | |
| forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish | |
| CHEBI:42471 | |
| CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O |
Specifications
| 66575-29-9 | |
| MFCD00082317 | |
| 14,2476 | |
| Soluble in ethanol,dimethylsulfoxide and chloroform; Insolule in water. | |
| CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O | |
| 410.507 | |
| CHEBI:42471 | |
| ≥98% |
| C22H34O7 | |
| 1692716 | |
| forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish | |
| OHCQJHSOBUTRHG-KGGHGJDLSA-N | |
| [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate | |
| 47936 | |
| 410.51 | |
| Forskolin |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P280-P302+P352-P312-P363-P501c
H312
missing translation for 'einecsNumber' : 266-410-9
missing translation for 'rtecsNumber' : QL6150000
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.