missing translation for 'onlineSavingsMsg'
Learn More

Food Red No.3 Aluminum Lake (Technical Grade), TRC

Product Code. 30392841
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392841 5 g -
30396880 10 g -
30401379 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392841 Supplier Toronto Research Chemicals Supplier No. TRCF6880055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12227-78-0
Molecular Formula C20 H6 I4 O5
Molecular Weight (g/mol) 833.876
Synonym 3’,6’-Dihydroxy-2',4',5',7'-tetraiodo-spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one Aluminum Salt, Food Red 14:1, Erythrosine Aluminum Lake
IUPAC Name 2',4',5',7'-tetraiodo-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-diolate
SMILES [O-]c1c(I)cc2c(Oc3c(I)c([O-])c(I)cc3C24OC(=O)c5ccccc45)c1I

Specifications

Quantity 5 g
InChI Formula InChI=1S/C20H8I4O5/c21-11-5-9-17(13(23)15(11)25)28-18-10(6-12(22)16(26)14(18)24)20(9)8-4-2-1-3-7(8)19(27)29-20/h1-6,25-26H/p-2
Formula Weight 833.64
Chemical Name or Material Food Red No.3 Aluminum Lake (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.