accessibility menu, dialog, popup

UserName

AlAlOIOIOIOOIOIOIOIOOIOIOIOIOOI

CAS RN 12227-78-0

IUPAC Name: dialuminium(3+) tris(1',3',6',8'-tetraiodo-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-2',7'-bis(olate))
Synonyms: dialuminium(3+) tris(1',3',6',8'-tetraiodo-3-oxospiro[2-benzofuran-1,9'-xanthene]-2',7'-bis(olate))
Molecular Weight (g/mol): 2555.61
Molecular Formula: C60H18Al2I12O15
InChi Key: VKPZSICYIGQEAN-UHFFFAOYSA-H
SMILES: [Al+3].[Al+3].[O-]C1=C(I)C=C2OC3=CC(I)=C([O-])C(I)=C3C3(OC(=O)C4=CC=CC=C34)C2=C1I.[O-]C1=C(I)C=C2OC3=CC(I)=C([O-])C(I)=C3C3(OC(=O)C4=CC=CC=C34)C2=C1I.[O-]C1=C(I)C=C2OC3=CC(I)=C([O-])C(I)=C3C3(OC(=O)C4=CC=CC=C34)C2=C1I
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results