missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folliculin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £395.00
Specifications
| Antigen | Folliculin |
|---|---|
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18231584
|
Novus Biologicals
NBP1-44995SS |
0.025 mL |
£169.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18718813
|
Novus Biologicals
NBP1-44995 |
0.1 mL |
£395.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Folliculin Polyclonal specifically detects Folliculin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Folliculin | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication | |
| Whole antisera with 0.1% Sodium Azide | |
| BHD skin lesion fibrofolliculoma protein, BHDFLCL, Birt-Hogg-Dube syndrome protein, DKFZp547A118, FLJ45004, FLJ99377, folliculin, MGC17998, MGC23445 | |
| FLCN | |
| IgG | |
| Unpurified | |
| This antibody is expected to detect all three isoforms of Folliculin protein ( UniProt # Q8NFG4) |
| Western Blot 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse | |
| Q8NFG4, Q8QZS3 | |
| 201163 | |
| N-terminal partial recombinant human Folliculin protein [Swiss-Prot# Q8NFG4] expressed in E. coli. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title