missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folliculin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-44995SS
This item is not returnable.
View return policy
Description
Folliculin Polyclonal specifically detects Folliculin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Folliculin | |
| Polyclonal | |
| Western Blot 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Q8NFG4, Q8QZS3 | |
| FLCN | |
| N-terminal partial recombinant human Folliculin protein [Swiss-Prot# Q8NFG4] expressed in E. coli. | |
| 0.025 mL | |
| Cancer, Cell Cycle and Replication | |
| 201163 | |
| Human, Mouse | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.1% Sodium Azide | |
| BHD skin lesion fibrofolliculoma protein, BHDFLCL, Birt-Hogg-Dube syndrome protein, DKFZp547A118, FLJ45004, FLJ99377, folliculin, MGC17998, MGC23445 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| This antibody is expected to detect all three isoforms of Folliculin protein ( UniProt # Q8NFG4) | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction