missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMRP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13755-1-AP-20UL
This item is not returnable.
View return policy
Description
Members of the Rho.Specifications
| FMRP | |
| Polyclonal | |
| Unconjugated | |
| FMR1 | |
| FMR1, FMRP, fragile X mental retardation 1, FRAXA, POF, POF1, Protein FMR 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14265, 2332, 24948 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35922, Q06787, Q80WE1 | |
| FMR1 | |
| FMRP Fusion Protein Ag4697 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction