missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMO3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-82415
This item is not returnable.
View return policy
Description
FMO3 Polyclonal specifically detects FMO3 in Mouse samples. It is validated for Western Blot.
Specifications
| FMO3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| dimethylaniline monooxygenase [N-oxide-forming] 3, Dimethylaniline oxidase 3, dJ127D3.1, EC 1.14.13.8, flavin containing monooxygenase 3, FMO 3, FMO form 2, FMO II, FMOII, Hepatic flavin-containing monooxygenase 3, hepatic flavin-containing monooxygenase-3, MGC34400, TMAU | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2328 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_032056 | |
| FMO3 | |
| Synthetic peptide towards Fmo3. Peptide sequence KPNVKEFTETSAVFEDGTMFEAIDCVIFATGYGYAYPFLDDSIIKSRNNE. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Equine: 100%; Human: 100%; Pig: 100%; Rabbit: 100%; Bovine: 92%; Canine: 92%; Mouse: 92%; Rat: 92%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction