missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | FMN1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231453
|
Novus Biologicals
NBP3-35318-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231967
|
Novus Biologicals
NBP3-35318-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FMN1 Polyclonal antibody specifically detects FMN1 in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| FMN1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| DKFZp686C2281, DKFZp686G2387, FLJ45135, FMNLimb deformity protein homolog, formin (limb deformity), formin 1, formin-1, LDDKFZP686C2281, MGC125288, MGC125289 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-495 of human FMN1 (NP_001096654.1).,, Sequence:, NIDMPKTEPKGADPESPRREEMGCNADQESQSGPGVPQTQGGEVKPKSPETALEAFKALFIRPPRKGTTADTSELEALKRKMRHEKESLRAVFERSNSKPADGPSDSKSPDHSLTEQDDRTPGRLQAVWPPPKTKDTEEKVGLKYT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 342184 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title