missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35318-20ul
This item is not returnable.
View return policy
Description
FMN1 Polyclonal antibody specifically detects FMN1 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| FMN1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| DKFZp686C2281, DKFZp686G2387, FLJ45135, FMNLimb deformity protein homolog, formin (limb deformity), formin 1, formin-1, LDDKFZP686C2281, MGC125288, MGC125289 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-495 of human FMN1 (NP_001096654.1).,, Sequence:, NIDMPKTEPKGADPESPRREEMGCNADQESQSGPGVPQTQGGEVKPKSPETALEAFKALFIRPPRKGTTADTSELEALKRKMRHEKESLRAVFERSNSKPADGPSDSKSPDHSLTEQDDRTPGRLQAVWPPPKTKDTEEKVGLKYT | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 342184 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction