missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CRHR2/CRF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30958-100ug
This item is not returnable.
View return policy
Description
CRHR2/CRF2 Polyclonal antibody specifically detects CRHR2/CRF2 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| CRHR2/CRF2 | |
| Polyclonal | |
| Western Blot : 1:400, ELISA, Immunohistochemistry-Paraffin : 2-10 μg/mL | |
| corticotropin releasing hormone receptor 2, corticotropin-releasing factor receptor 2, Corticotropin-releasing hormone receptor 2, CRF2, CRF2R, CRFR2, CRF-R2, CRF-R-2, CRFR-2, CRH2R, CRH-R2, CRH-R-2 | |
| Synthetic peptide derived from the N-terminal region of the CRHR2/CRF2 protein. | |
| 100 μg | |
| GPCR, Neuroscience, Neurotransmission | |
| 1395 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction