missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CRHR2/CRF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£162.00 - £386.00

Specifications

Antigen CRHR2/CRF2
Dilution Western Blot : 1:400, ELISA, Immunohistochemistry-Paraffin : 2-10 μg/mL
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30066848
View Documents
Novus Biologicals™
NBP3-30958-100ug
100 μg
£386.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30067170
View Documents
Novus Biologicals™
NBP3-30958-25ug
25 μg
£162.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CRHR2/CRF2 Polyclonal antibody specifically detects CRHR2/CRF2 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications

Specifications

CRHR2/CRF2
ELISA, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
GPCR, Neuroscience, Neurotransmission
Phosphate buffered saline
1395
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Western Blot : 1:400, ELISA, Immunohistochemistry-Paraffin : 2-10 μg/mL
Polyclonal
Purified
RUO
Human
corticotropin releasing hormone receptor 2, corticotropin-releasing factor receptor 2, Corticotropin-releasing hormone receptor 2, CRF2, CRF2R, CRFR2, CRF-R2, CRF-R-2, CRFR-2, CRH2R, CRH-R2, CRH-R-2
Synthetic peptide derived from the N-terminal region of the CRHR2/CRF2 protein.
Antigen Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.