missing translation for 'onlineSavingsMsg'
Learn More

Fluphenazine N4'-oxide, TRC

Product Code. 30394294
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394294 25 mg
30405169 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394294 Supplier Toronto Research Chemicals Supplier No. TRCF59842025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30897-25-7
Molecular Formula C22H26F3N3O2S
Molecular Weight (g/mol) 453.52
Synonym 4-[3-[2-(Trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-1-piperazineethanol 1-Oxide
SMILES IC1=C(O)C(I)=CC(O[13C]2=[13C](I)[13CH]=[13C]([13CH2][13C@@H]([13C](O)=O)[15NH2])[13CH]=[13C]2I)=C1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H26F3N3O2S/c23-22(24,25)17-6-7-21-19(16-17)27(18-4-1-2-5-20(18)31-21)9-3-8-26-10-12-28(30,13-11-26)14-15-29/h1-2,4-7,16,29H,3,8-15H2
Formula Weight 453.1698
Chemical Name or Material Fluphenazine N4'-oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.