CAS RN 30897-25-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fluphenazine N4'-oxide, TRC
CAS: 30897-25-7 Molecular Formula: C22H26F3N3O2S Molecular Weight (g/mol): 453.52 Synonym: 4-[3-[2-(Trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-1-piperazineethanol 1-Oxide SMILES: IC1=C(O)C(I)=CC(O[13C]2=[13C](I)[13CH]=[13C]([13CH2][13C@@H]([13C](O)=O)[15NH2])[13CH]=[13C]2I)=C1