missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) FluoroTrans™ W PVDF Transfer Membrane

FluoroTrans W membrane is optimized for Western transfer applications.

Brand:  Cytiva (formerly Pall Lab) BSP0157

Additional Details : Weight : 0.18050kg

 View more versions of this product

Product Code. 17184711

  • £206.00 / Pack of 10
Estimated Shipment: 30-10-2024
to see stock.
Add to basket

Description

Description

  • This membrane allows for sensitive protein detection with low background and very low protein burn-through
  • FluoroTrans W membrane is compatible with all total protein stains and immunodetection methods
Specifications

Specifications

Sheets
FluoroTrans PVDF
10 x 15 cm
Non-sterile
0.2 μm
127 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers