Learn More
Thermo Scientific™ Fluoroskan™ FL Microplate Fluorometer and Luminometer, two dispensers
The Thermo Scientific Fluoroskan FL is a compact and robust microplate fluorometer and luminometer, offering simplicity and reliability for increased efficiency of workflows.
Brand: Thermo Scientific™ 5200222
Description
Featuring excellent optical performance, the Fluoroskan FL Microplate Fluorometer and Luminometer is ideal for life science fluorometric and luminometric assays. The Fluoroskan FL fluorometer and luminometer offers plate format versatility, fast plate reading speeds, and up to three dispensers.
• High sensitivity for both top and bottom reading
• Fast reading speed essential for kinetic applications
• Onboard dispensers for precise initiation of kinetic reactions
• Powerful and easy-to-use Thermo Scientific SkanIt software
High sensitivity for both top and bottom reading
The use of fiberless direct illumination optics for both top and bottom reading enables high sensitivity, a wide dynamic range, and accurate and precise results for 6- to 384-well plates.
Fast reading speed essential for kinetic applications
The Fluoroskan FL Microplate Fluorometer and Luminometer is capable of reading a 96-well plate in just 18 seconds, essential for kinetic applications. Fluorometric GFP measurement and luminometric luciferase measurement from the same well occurs quickly due to the versatility of the assay programming.
Onboard dispensers precise initiation of kinetic reactions
The Fluoroskan FL Microplate Fluorometer and Luminometer can be equipped with up to three reagent dispensers* for use during fast kinetic assays, such as Ca2+ flux assays. The onboard dispensers support simultaneous dispensing and reading, enabling the monitoring of fast kinetic measurements from the very start of the reaction. Very low dead volume and backflush capability support conservation of reagents.
*Upgrade to third dispenser must be performed by service technician
Powerful and easy-to-use SkanIt software
With a visual workflow setup, SkanIt software is quick and easy to learn. It offers effortless data analysis (with a comprehensive selection of built-in calculations, including blank subtraction, standard curve creation, classification, quality control, kinetic analysis) and exporting capabilities. Each user can install SkanIt software on his/her own computer to set up assays and analyze data.
Includes:
SkanIt PC Software and the following filter pairs (excitation/emission): 355 nm/460 nm, 485 nm/538 nm. An extensive selection of additional filters is available upon request.
Applications include:
Ca2+ flux assays , cell proliferation, cytotoxicity, cell adhesion, nucleic acid quantitation, reporter gene assays, hybridization assays, ion channel, FRET assays, BRET assays, immunoassays, enzyme activity, bacterial quantitation, phagocytosis, oligonucleotide assays
Specifications
Robot | |
420 x 420 x 340 mm (16.5 x 16.5 x 13.4 in.) | |
PC Software (Thermo Scientific SkanIt Software) | |
18 s for 96-well Plate | |
6 to 384-well Plates | |
Top and Bottom | |
Orbital | |
Filters | |
Room Temperature |
Photomultiplier Tube (PMT) | |
Ambient +3°C to 45°C | |
Quartz Halogen Lamp | |
Fluorescence Intensity, Luminescence | |
Fluoroskan | |
Up to approx. 5000 Relative Fluorescence Units (RFU) | |
SkanIt Software | |
200 VA max. |