missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Fluorescence Test Plate for Synergy Neo2/H1/2/HTX, FLx800 and Cytation Multi-Mode Reader
Shop All Agilent BioTek Products

Click to view available options
For Use With (Equipment):
Synergy Neo2/H1/2/HTX, FLx800 and Cytation Multi-Mode Readers
Unit Size:
Each
Description
Includes
Certificate of calibration, Flash drive with user manual and test protocols, Dust blower and cleaning swabs
- Designed to replace fluorescent liquid testing to check instrument performance
- Solid test plate
- Immune to the pipetting errors and evaporation issues
- Convenient size and multi-parameter functionality
- Checks for fluorescence intensity linearity-green (excitation 485nm, emission 528nm), blue (excitation 360nm, emission 460nm), Carrier flatness, Fluorescence polarization and Time-resolved fluorescence
Specifications
Specifications
| Product Type | Fluorescence Test Plate |
| For Use With (Equipment) | Synergy Neo2/H1/2/HTX, FLx800 and Cytation Multi-Mode Readers |
| Excitation/Emission | EX 485 nm, EM 528 nm |
| For Use With (Application) | Fluorescence |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction