missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Fluorescein Diacetate

Product Code. 16401178
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401178 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401178 Supplier MedChemExpress Supplier No. HYD0719
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fluorescein diacetate is a cell permeable esterase-substrate. Fluorescein diacetate can be used as a fluorogenic substrate for hGSTP1-1.

Specifications

Quantity 5 g
Product Type Biochemical Assay Reagents
Molecular Weight (g/mol) 416.38 g/mol
Solubility DMSO : 26.67 mg/mL (64.05 mM; Need ultrasonic)
Content And Storage -20°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
Target HBV
CAS 596-09-8
Synonym 3,6-Diacetoxyfluoran ∣ Di-O-acetylfluorescein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.