missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein biotin, 5-((N-(5-(N-(6-(biotinoyl)amino)hexanoyl)amino)pentyl)thioureidyl)fluorescein

Product Code. 11584147
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11584147

Brand: Invitrogen™ B1370

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

5-((N-(5-(N-(6-(biotinoyl)amino)hexanoyl)amino)pentyl)thioureidyl)fluorescein (fluorescein biotin)

Fluorescein biotin was developed as an alternative to radioactive biotin for detecting and quantitating biotin-binding sites by either fluorescence or absorbance. Upon binding to avidin or streptavidin, the fluorescence or absorbance of this molecule is quenched.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type Molecular Probes Fluorescein biotin
Content And Storage Store at room temperature and protect from light.
Shipping Condition Room Temperature
Label or Dye FITC (Fluorescein)

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.