missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein, Armenian Hamster anti-Non-species specific, Clone: 4-4-20 (enhanced), Novus Biologicals™
Armenian Hamster Monoclonal Antibody
£198.00 - £510.00
Specifications
| Antigen | Fluorescein |
|---|---|
| Clone | 4-4-20 (enhanced) |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Flow Cytometry, Control |
| Applications | Western Blot, Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18286379
|
Novus Biologicals
NBP2-81025-0.025MG |
0.025 mg |
£198.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18284169
|
Novus Biologicals
NBP2-81025-0.2MG |
0.2 mg |
£510.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fluorescein Monoclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, Control.Specifications
| Fluorescein | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Armenian Hamster | |
| Non-species specific | |
| FITC, fluorescein isothyocianate | |
| IgG κ | |
| Protein A purified | |
| Fluorescein labelled proteins. |
| 4-4-20 (enhanced) | |
| Western Blot, Flow Cytometry, Control | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| Fluorescein I-bovine serum albumin (FlI-BSA). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title