missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein, Armenian Hamster anti-Non-species specific, Clone: 4-4-20 (enhanced), Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP2-81025-0.2MG
This item is not returnable.
View return policy
Description
Fluorescein Monoclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, Control.
Specifications
| Fluorescein | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Control | |
| FITC, fluorescein isothyocianate | |
| Fluorescein I-bovine serum albumin (FlI-BSA). | |
| 0.2 mg | |
| Primary | |
| Non-species specific | |
| Purified |
| Western Blot, Flow Cytometry | |
| 4-4-20 (enhanced) | |
| Unconjugated | |
| PBS | |
| Armenian Hamster | |
| Protein A purified | |
| RUO | |
| Fluorescein labelled proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction