missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Fluorescein-5-maleimide

Product Code. 16498748
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498748 10 mg 10mg
16464667 50 mg 50mg
16453816 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498748 Supplier MedChemExpress Supplier No. HYD0098
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fluorescein-5-maleimide is a fluorescent thiol-reactive dye used to conjugate fluorescein to proteins (excitation: 494 nm, emission: 519 nm).

Specifications

Quantity 10 mg
Product Type Dye Reagents
Molecular Weight (g/mol) 427.36 g/mol
Solubility DMSO : 50 mg/mL (117.00 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble)
Content And Storage -20°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
Target HBV
CAS 75350-46-8
Synonym N-(5-Fluoresceinyl)maleimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.