missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ FluoReporter™ FITC Protein Labeling Kit

Product Code. 10195082
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10195082 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10195082 Supplier Molecular Probes™ Supplier No. F6434

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The FluoReporter™ FITC Protein Labeling Kit provides a convenient and flexible means for attaching fluorescein to your monoclonal or polyclonal antibody (or any protein >30 kDa).

The FluoReporter™ FITC Protein Labeling Kit provides a convenient and flexible means for attaching fluorescein (FITC, Ex/Em ∼494/518 nm) to your monoclonal or polyclonal antibody (or any protein >30 kDa). The kit supplies the reagents needed for 5 to 10 labeling and separation reactions.

Important Features of the FluoReporter™ FITC Protein Labeling Kit:
• Labeled proteins typically ready to use in under 3 hours with very little hands-on time
• Optimized for 0.2–2.0 mg of protein with molecular weight >30 kDa
• Labeled proteins are purified using convenient spin filters
• Requires the antibody or protein be purified prior to labeling

Learn More About Protein Labeling
We offer a wide selection of Molecular Probes™ protein and antibody labeling kits to fit your starting material and your experimental setup. See Antibody Labeling from A to Z or use our Labeling Chemistry Selection Tool for other choices. To learn more about our various kits read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in the Molecular Probes™ Handbook.

We’ll Make a Custom Labeled Protein for You
If you can’t find what you’re looking for in our stocked list, we’ll prepare a custom labeled protein for you. Our custom services are efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Labeling Kit
Quantity 1 kit
Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Detection Method Fluorescence
Labeling Scale 0.2 to 2 mg
Shipping Condition Room Temperature
Product Line FluoReporter
Label or Dye FITC (Fluorescein)
Labeling Target Antibodies, Proteins

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.