missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Fluo-4FF, AM, cell permeant - Special Packaging

Product Code. 11564806
Click to view available options
Quantity:
10 x 50 μg
Unit Size:
500µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11564806

Brand: Molecular Probes™ F23981

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Fluo-4FF, AM, cell permeant - Special Packaging

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. Fluo-5F, fluo-5N, and fluo-4ff are analogs of fluo-4 with lower Ca2+-binding affinity, making them suitable for detecting intracellular calcium levels in the 1μM to 1 mM range that would saturate the response of fluo-3 and fluo-4. Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells. These indicators are compatible with excitation at 488 nm by argon-ion laser sources, making them useful for confocal microscopy, flow cytometry, and microplate screening applications.

Calcium Indicator (AM Ester) Specifications:
  • Label (Ex/Em of Ca2+–bound form): Fluo-4ff (494/516 nm)
  • Fluorescence intensity increase upon binding Ca2+: >100 fold
  • Kd for Ca2+ in buffer: ∼9.7μM
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength

    Using TPEN to Control Heavy Metal Cations
    In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator TPEN.

    More Choices for Fluorescent Calcium Indicators
    We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.

    For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.

    For Research Use Only. Not for human or animal therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store in freezer -5°C to -30°C and protect from light.
    Detection Method Fluorescence
    For Use With (Application) Cell Viability and Proliferation
    For Use With (Equipment) Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager
    Product Type Dye
    Dye Type Fluorescent Dye-Based
    Quantity 10 x 50 μg
    Shipping Condition Room Temperature

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.