missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Fluo-4 NW Calcium Assay Kit

Product Code. p-7075901
Click to view available options
Quantity:
10 Microplates
100 Microplate(s)
Unit Size:
1 set
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10266762

Brand: Molecular Probes™ F36206

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Fluo-4 NW Calcium Assay Kit

*The Fluo-4 NW (no-wash) Calcium Assay Kits provide a simplified method for detecting intracellular calcium, without a quencher dye or a wash step.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Calcium Assay
For Use With (Equipment) Microplate Reader
Product Type Stain
Dye Type Fluorescent Dye-Based
Quantity 10 Microplates
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.