missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Fluo-4 AM - Packaged For High-Throughput Screening
Click to view available options
Quantity:
5 x 1 mg
Unit Size:
5mg
Description
Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. Fluo-3 has been used to image the spatial dynamics of Ca2+ signaling, in flow cytometry experiments involving photoactivation of caged chelators, second messengers, and neurotransmitters, and for cell-based pharmacological screening. Fluo-4 is an analog of fluo-3 with the two chlorine substituents replaced by fluorines, which results in increased fluorescence excitation at 488 nm and consequently higher fluorescence signal levels. Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells. These indicators are useful for fluorescence and confocal microscopy, flow cytometry, and microplate screening applications.
Calcium Indicator (AM Ester) Specifications:
Label (Ex/Em of Ca2+–bound form): Fluo-4 (494/506 nm)
Fluorescence intensity increase upon binding Ca2+: >100 fold
Kd for Ca2+ in buffer: ∼335 nM
Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
Using TPEN to Control Heavy Metal Cations
In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator TPEN.
More Choices for Fluorescent Calcium Indicators
We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.
For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Calcium Indicator (AM Ester) Specifications:
Using TPEN to Control Heavy Metal Cations
In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator TPEN.
More Choices for Fluorescent Calcium Indicators
We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.
For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
Specifications
| Content And Storage | Store at -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Cell Viability, Proliferation and Cellular Imaging |
| For Use With (Equipment) | Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager |
| Product Type | Dye |
| Dye Type | Fluorescent Dye-Based |
| Quantity | 5 x 1 mg |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction