missing translation for 'onlineSavingsMsg'
Learn More

Flumequine, 98%, Thermo Scientific™

Inhibits topoisomerases in gram negative bacteria

£32.70 - £94.70

Chemical Identifiers

CAS 42835-25-6
Molecular Formula C14H12FNO3
Molecular Weight (g/mol) 261.252
MDL Number MFCD00079298
InChI Key DPSPPJIUMHPXMA-UHFFFAOYSA-N
Synonym 9-Fluoro-1,5,6,7-tetrahydro-5-methyl-1-oxopyrido[3,2,1-ij]quinoline-2-carboxylic Acid
PubChem CID 3374
ChEBI CHEBI:85269
SMILES CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2)F)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15497639
SDS View Documents
Thermo Scientific Alfa Aesar
J66332.03
1g
£32.70
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15407649
SDS View Documents
Thermo Scientific Alfa Aesar
J66332.06
5g
£94.70
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

42835-25-6
261.252
DPSPPJIUMHPXMA-UHFFFAOYSA-N
3374
CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2)F)C(=O)O
C14H12FNO3
MFCD00079298
9-Fluoro-1,5,6,7-tetrahydro-5-methyl-1-oxopyrido[3,2,1-ij]quinoline-2-carboxylic Acid
CHEBI:85269

Specifications

42835-25-6
>110°C (230°F)
MFCD00079298
14,4137
Soluble in DMSO and dilute alkali hydroxides; Insoluble in water
CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2)F)C(=O)O
3374
261.25
Flumequine
White
C14H12FNO3
490724
9-Fluoro-1,5,6,7-tetrahydro-5-methyl-1-oxopyrido[3,2,1-ij]quinoline-2-carboxylic Acid
DPSPPJIUMHPXMA-UHFFFAOYSA-N
261.252
CHEBI:85269
98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 255-962-6

missing translation for 'rtecsNumber' : DK1672000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.