missing translation for 'onlineSavingsMsg'
Learn More

Flumequine, 98%, Thermo Scientific™

Inhibits topoisomerases in gram negative bacteria

Brand:  Thermo Scientific Alfa Aesar J66332.06

Quantity 5g

 View more versions of this product

Product Code. 15407649

  • £94.70 / 5g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

42835-25-6
261.252
DPSPPJIUMHPXMA-UHFFFAOYSA-N
3374
CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2)F)C(=O)O
C14H12FNO3
MFCD00079298
9-Fluoro-1,5,6,7-tetrahydro-5-methyl-1-oxopyrido[3,2,1-ij]quinoline-2-carboxylic Acid
CHEBI:85269

Specifications

42835-25-6
5g
C14H12FNO3
490724
9-Fluoro-1,5,6,7-tetrahydro-5-methyl-1-oxopyrido[3,2,1-ij]quinoline-2-carboxylic Acid
DPSPPJIUMHPXMA-UHFFFAOYSA-N
261.252
CHEBI:85269
98%
White
>110°C (230°F)
MFCD00079298
14,4137
Soluble in DMSO and dilute alkali hydroxides; Insoluble in water
CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2)F)C(=O)O
3374
261.25
Flumequine
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.