missing translation for 'onlineSavingsMsg'
Learn More

Fluconazole, 99%, Thermo Scientific™

Fluconazole, CAS # 86386-73-4, is a synthetic triazole with antifungal activity against some species of Candida. It also shows inhibitory activity against cytochrome P-450 enzymes and it is a demethylase.

Brand:  Thermo Scientific Alfa Aesar J62015.06

Quantity 5g

 View more versions of this product

Product Code. 15464479

  • ONLINE EXCLUSIVE PRICE
    £257.00
    £231.65 / 5g
    Save £25.35
    9% Off
 Restricted Item
Estimated Shipment: 07-10-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Fluconazole, 98% is a triazole compound showing antifungal activity mainly against Candida, Coccidioides, and Cryptococcus species
• It has been shown to inhibit fungal cytochrome P-450 sterol C-14 α-demethylation, resulting in the accumulation of fungal 14 α-methyl sterols, the loss of normal fungal sterols, and associated fungistatic activity
• It also inhibits cytochrome P450 family 2 subfamily c member 9 (CYP2C9)
• Mammalian cell demethylation is much less susceptible to fluconazole inhibition
Applications
• Fluconazole has fungistatic activity against many fungal strains including Candida albicans, Candida glabrata, Candida parapsilosis, Candida tropicalis, and Cryptococcus neoformans.
• Fluconazole interferes with fungal ergosterol synthesis and downregulates the metallothionein gene
• In biological experiments, this compound has been used to assess minimum inhibitory concentration in (MIC)-fungal assays
• It has also been used as a positive control in antifungal assay of plants
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

86386-73-4
306.277
RFHAOTPXVQNOHP-UHFFFAOYSA-N
3365
2-(2,4-difluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol
C13H12F2N6O
MFCD00274549
fluconazole, diflucan, triflucan, elazor, biozolene, biocanol, fungata, fluconazol, fluconazolum, flucazol
CHEBI:46081
C1=CC(=C(C=C1F)F)C(CN2C=NC=N2)(CN3C=NC=N3)O

Specifications

86386-73-4
C13H12F2N6O
5g
fluconazole, diflucan, triflucan, elazor, biozolene, biocanol, fungata, fluconazol, fluconazolum, flucazol
RFHAOTPXVQNOHP-UHFFFAOYSA-N
2-(2,4-difluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol
3365
306.27
Fluconazole
1.05
MFCD00274549
14,4122
Soluble indimethylsulfoxide and ethanol.
C1=CC(=C(C=C1F)F)C(CN2C=NC=N2)(CN3C=NC=N3)O
306.277
CHEBI:46081
99%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.