missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flt-3/Flk-2/CD135 Antibody (MM0294-9G23) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£171.00 - £424.00
Specifications
| Antigen | Flt-3/Flk-2/CD135 |
|---|---|
| Clone | MM0294-9G23 |
| Concentration | LYOPH |
| Applications | CyTOF, Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18055887
|
Novus Biologicals
NBP2-12311-0.025MG |
0.025 mg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18715563
|
Novus Biologicals
NBP2-12311 |
0.1 mg | |||||||
Description
Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Human samples. It is validated for Western Blot.Specifications
| Flt-3/Flk-2/CD135 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Cynomolgus Monkey, Human | |
| CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
| FLT3 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0294-9G23 | |
| CyTOF, Western Blot, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| P36888 | |
| 2322 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title