missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flt-3/Flk-2/CD135 Antibody (MM0294-9G23) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12311-0.025MG
This item is not returnable.
View return policy
Description
Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Human samples. It is validated for Western Blot.
Specifications
| Flt-3/Flk-2/CD135 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1000, Flow Cytometry, CyTOF-ready | |
| P36888 | |
| FLT3 | |
| Human Flt3/CD135 | |
| 0.025 mg | |
| Cell Cycle and Replication | |
| 2322 | |
| Cynomolgus Monkey, Human | |
| Purified |
| CyTOF, Western Blot, Flow Cytometry | |
| MM0294-9G23 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction