missing translation for 'onlineSavingsMsg'
Learn More

De Buyer™ White Polypropylene Flour Scoop

Product Code. 11922845
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11922845 1000 mL Each
11947844 250 mL Each
11957844 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11922845 Supplier De Buyer™ Supplier No. 4370.09
Add to basket
Add to basket
This item is not returnable. View return policy

Pour dry goods with the De Buyer™ White Polypropylene Flour Scoop, molded in one piece from polypropylene and resulting in a robust single-piece design that is dishwasher safe.

  • Handle molded in PP:and inseparable from the rest of the scoop
  • Single-piece design
  • Dishwasher-safe

Specifications

Capacity (Metric) 1000 mL
Color White
Product Type Flour Scoop
Material Polypropylene
Handle Type Molded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.