missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flotillin 1 Mouse anti-Human, Unconjugated, Clone: 1D6C6, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Flotillin 1 |
|---|---|
| Clone | 1D6C6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17521486
|
Proteintech
67968-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17511486
|
Proteintech
67968-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Flotillin 1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human Flotillin 1. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The Flotillin 1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Flotillin 1 | |
| Western Blot | |
| Unconjugated | |
| Human, Mouse, Rat, Pig, Rabbit | |
| IgG2b |
| 1D6C6 | |
| Monoclonal | |
| Mouse | |
| 10211 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title