Learn More
Flotillin 1 Mouse anti-Human, Unconjugated, Clone: 1D6C6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67968-1-IG-150UL
Description
The Flotillin 1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human Flotillin 1. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The Flotillin 1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Flotillin 1 | |
| Monoclonal | |
| Unconjugated | |
| 150 μL | |
| 10211 | |
| IgG2b |
| Western Blot | |
| 1D6C6 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig, Rabbit |