missing translation for 'onlineSavingsMsg'
Learn More

FLJ10808 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

£158.00 - £302.00

Specifications

Antigen FLJ10808
Concentration 0.5 mg/mL
Dilution Western Blot 0.5-2.0 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18170802
View Documents
Novus Biologicals
NBP2-24534SS
0.025 mg
£158.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18166065
View Documents
Novus Biologicals
NBP2-24534
0.1 mg
£302.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FLJ10808 Polyclonal specifically detects FLJ10808 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

FLJ10808
Western Blot 0.5-2.0 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Polyclonal
Rabbit
Human
A0AVT1
55236
A portion of amino acids 775-825 of human UBE1L2 was used as the immunogen for this antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
RUO
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
E1-L2, FLJ10808, FLJ23367, Monocyte protein 4, MOP4, MOP-4, UBA6, ubiquitin-activating enzyme E1, UBE1L2, Ubiquitin-activating enzyme 6, ubiquitin-activating enzyme E1-like 2, Ubiquitin-activating enzyme E1-like protein 2, ubiquitin-like modifier activating enzyme 6, ubiquitin-like modifier-activating enzyme 6
UBA6
IgG
Affinity Purified
The amino acid sequence used as immunogen for this antibody is 100% homologous in human (isoforms CRA_a and CRA_b) and 88% homologous in monkey.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.