missing translation for 'onlineSavingsMsg'
Learn More

FLJ10808 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

Brand:  Novus Biologicals NBP2-24534

 View more versions of this product

Product Code. 18166065

  • £302.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FLJ10808 Polyclonal specifically detects FLJ10808 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FLJ10808
Polyclonal
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
E1-L2, FLJ10808, FLJ23367, Monocyte protein 4, MOP4, MOP-4, UBA6, ubiquitin-activating enzyme E1, UBE1L2, Ubiquitin-activating enzyme 6, ubiquitin-activating enzyme E1-like 2, Ubiquitin-activating enzyme E1-like protein 2, ubiquitin-like modifier activating enzyme 6, ubiquitin-like modifier-activating enzyme 6
Rabbit
Affinity Purified
RUO
55236
Human
IgG
Western Blot, Immunohistochemistry (Paraffin)
0.5 mg/mL
Western Blot 0.5-2.0 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
A0AVT1
UBA6
A portion of amino acids 775-825 of human UBE1L2 was used as the immunogen for this antibody.
0.1 mg
Primary
The amino acid sequence used as immunogen for this antibody is 100% homologous in human (isoforms CRA_a and CRA_b) and 88% homologous in monkey.
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only