Learn More
Proteintech FlexAble CoraLite Plus 647 Antibody Labeling Kit for Human IgG
Product Code. p-200050231 Shop All Proteintech Products
Click to view available options
Quantity:
10 Tests
100 Tests
50 Tests
Unit Size:
10 reactions
200 reactions
50 reactions
Product Code. 18039345
Brand: Proteintech undefined
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
FlexAble CoraLite Plus 647 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite Plus 647 in any buffer condition to Human IgG primary antibodies from any supplier.
FlexAble CoraLite™ Plus 647 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 647 in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Specifications
Quantity 50 Tests
Format Liquid