missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ FlexAble CoraLite™ Plus 550 Antibody Labeling Kit for Rabbit IgG

Product Code. 17781509 Shop All Proteintech Products
Click to view available options
Quantity:
10 x 0.5 μg Reactions
200 x 0.5 μg Reactions
50 x 0.5 μg Reactions
Unit Size:
10 reactions
200 reactions
50 reactions
This item is not returnable. View return policy

Product Code. 17781509

Brand: Proteintech™ KFA002200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Antibody Labeling Kit conjugates CoraLite™ 550 into Rabbit IgG antibodies.

FlexAble CoraLite™ Plus 550 Antibody Labeling Kit for Rabbit IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 550 in any buffer condition to rabbit IgG primary antibodies from any supplier.

  • One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
  • Label your rabbit IgG antibody in <10 min without any additional equipment

Specifications

Kit Contents 4 x 50 μL CoraLite™ 550 FlexLinker for Rabbit IgG, 4 x 100 μL FlexQuencher for Rabbit IgG, 4 x 500 μL FlexBuffer, Manual, Protocol Magnet
Quantity 200 x 0.5 μg Reactions
Content And Storage 1 Year at -20°C or 6 Months at +4°C
Excitation/Emission 554 nm/570 nm
Labeling Target IgG
Species Rabbit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.