missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLASH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77301-0.025mg
This item is not returnable.
View return policy
Description
FLASH Polyclonal specifically detects FLASH in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| FLASH | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CASP8 associated protein 2, caspase 8 associated protein 2, CED-4, FLASHKIAA1315CASP8-associated protein 2, FLICE associated huge, FLICE-associated huge protein, FLJ11208, human FLASH, RIP25FLASH homolog RIP25 | |
| Rabbit | |
| 220 kDa | |
| 0.025 mg | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 9994 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL | |
| AF165161 | |
| CASP8AP2 | |
| Antibody was raised against a synthetic peptide corresponding to amino acids near the carboxy terminus of human FLASH which differ from those of mouse by one amino acid. The immunogen is located within the last 50 amino acids of FLASH. Amino Acid Squence: SERFQQLMKLFEKSKC | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction