missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLASH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£133.00
Specifications
| Antigen | FLASH |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
FLASH Polyclonal specifically detects FLASH in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| FLASH | |
| Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| PBS with 0.02% Sodium Azide | |
| CASP8 associated protein 2, caspase 8 associated protein 2, CED-4, FLASHKIAA1315CASP8-associated protein 2, FLICE associated huge, FLICE-associated huge protein, FLJ11208, human FLASH, RIP25FLASH homolog RIP25 | |
| CASP8AP2 | |
| IgG | |
| Affinity Purified | |
| 220 kDa |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| AF165161 | |
| 9994 | |
| Antibody was raised against a synthetic peptide corresponding to amino acids near the carboxy terminus of human FLASH which differ from those of mouse by one amino acid. The immunogen is located within the last 50 amino acids of FLASH. Amino Acid Squence: SERFQQLMKLFEKSKC | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title