missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FLAP Recombinant Rabbit Monoclonal Antibody (23GB3940), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | FLAP |
|---|---|
| Clone | 23GB3940 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089915
![]() |
Invitrogen™
MA552644 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088275
|
Invitrogen™
MA552645 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FLAP Recombinant Monoclonal antibody specifically detects FLAP in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| FLAP | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P20292 | |
| 241 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3940 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 5-lipoxygenase-activating protein FLAP; Alox5ap; arachidonate 5-lipoxygenase activating protein; arachidonate 5-lipoxygenase-activating protein; Flap; MK-886-binding protein | |
| A synthesized peptide derived from FLAP (130-161AA). | |
| Primary | |
| -20°C | |
| ALOX5AP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title