missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FLAP Recombinant Rabbit Monoclonal Antibody (23GB3940), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552645
This item is not returnable.
View return policy
Description
ALOX5AP is a protein which, with 5-lipoxygenase, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.
Specifications
| FLAP | |
| Recombinant Monoclonal | |
| 0.58 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P20292 | |
| ALOX5AP | |
| A synthesized peptide derived from FLAP (130-161AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3940 | |
| Unconjugated | |
| ALOX5AP | |
| 5-lipoxygenase-activating protein FLAP; Alox5ap; arachidonate 5-lipoxygenase activating protein; arachidonate 5-lipoxygenase-activating protein; Flap; MK-886-binding protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 241 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction