missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP51/FKBP5 Antibody (Hi51B), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£194.00
Specifications
| Antigen | FKBP51/FKBP5 |
|---|---|
| Clone | Hi51B |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Western Blot |
Description
FKBP51/FKBP5 Monoclonal specifically detects FKBP51/FKBP5 in Human, Mouse, Rat, Canine, Hamster, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| FKBP51/FKBP5 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| 51 kDa FKBP, 54 kDa progesterone receptor-associated immunophilin, AIG6,51 kDa FK506-binding protein, Androgen-regulated protein 6, FF1 antigen, FK506 binding protein 5, FK506-binding protein 5, FKBP-5, FKBP-51, FKBP51PPIase FKBP5, FKBP54EC 5.2.1.8, HSP90-binding immunophilin, MGC111006, p54, P54,51 kDa FK506-binding protein 5, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase FKBP5, PPIase, Ptg-10, rotamase, T-cell FK506-binding protein | |
| FKBP5 | |
| IgG1 | |
| Affinity Purified | |
| Detects approx 51kDa. |
| Hi51B | |
| Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Monoclonal | |
| Mouse | |
| Cancer | |
| Q13451 | |
| 2289 | |
| Synthetic peptide corresponding to the residues of human FKBP51 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title