missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP51/FKBP5 Antibody (Hi51B), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB110-96873-0.025mg
This item is not returnable.
View return policy
Description
FKBP51/FKBP5 Monoclonal specifically detects FKBP51/FKBP5 in Human, Mouse, Rat, Canine, Hamster, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| FKBP51/FKBP5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| 51 kDa FKBP, 54 kDa progesterone receptor-associated immunophilin, AIG6,51 kDa FK506-binding protein, Androgen-regulated protein 6, FF1 antigen, FK506 binding protein 5, FK506-binding protein 5, FKBP-5, FKBP-51, FKBP51PPIase FKBP5, FKBP54EC 5.2.1.8, HSP90-binding immunophilin, MGC111006, p54, P54,51 kDa FK506-binding protein 5, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase FKBP5, PPIase, Ptg-10, rotamase, T-cell FK506-binding protein | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects approx 51kDa. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| Hi51B | |
| Unconjugated | |
| Q13451 | |
| FKBP5 | |
| Synthetic peptide corresponding to the residues of human FKBP51 | |
| 0.025 mg | |
| Cancer | |
| 2289 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction