missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP12 Rabbit anti-Human, Mouse, Rat, Clone: 9A2J5, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | FKBP12 |
|---|---|
| Clone | 9A2J5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18336433
|
Bio-Techne
NBP3-15418-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18368186
|
Novus Biologicals
NBP3-15418-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FKBP12 Monoclonal antibody specifically detects FKBP12 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| FKBP12 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, mTOR Pathway | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2280 | |
| IgG | |
| Affinity purified |
| 9A2J5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase | |
| A synthetic peptide corresponding to a sequence within amino acids 29-108 of human FKBP12 (P62942). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title