missing translation for 'onlineSavingsMsg'
Learn More

FKBP12 Rabbit anti-Human, Mouse, Rat, Clone: 9A2J5, Novus Biologicals™

Product Code. 18336433 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336433 20 μg 20µL
18368186 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18336433 Supplier Bio-Techne Supplier No. NBP31541820UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FKBP12 Monoclonal antibody specifically detects FKBP12 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP12
Applications Western Blot
Classification Monoclonal
Clone 9A2J5
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 29-108 of human FKBP12 (P62942).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Autophagy, Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2280
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.