Learn More
Trajan™ Fixed-Needle Autosampler Syringes for Thermo Scientific Instruments
For reduced non specific sample interaction and increased analyte recovery
Brand:  Trajan™ 0029741
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 16672572
  • £59.00 / Each
Estimated Shipment: 07-06-2024
to see stock.
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Specifications
22 G
0.72 mm
3.14 in.
10F-BT-8/22C
10 μL
Thermo AS2000
0.02 in.
Gas chromatography
Autosampler Syringe
Ea.
Cone
Fixed