missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Fixed-Needle Autosampler Syringes for Thermo Scientific Instruments

For reduced non specific sample interaction and increased analyte recovery

Brand:  Trajan™ 0029761

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15218463

  • £287.00 / Pack of 6
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications

Specifications

26 G
0.47 mm
1.96 in.
10F-BT-GT-5/0.47C
10 μL
Thermo Scientific instruments AS3000, AS2000, AS200/800
0.018 in.
Gas chromatography
Autosampler Syringe
6 Pk.
Cone
Fixed
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers