missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Fixable Viability Dye eFluor™ 780

Product Code. 13539140
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
Unit Size:
100 tests
500 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13539140 100 Tests 100 tests
15383562 500 Tests 500 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13539140 Supplier Invitrogen™ Supplier No. 65086514
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

A viability dye that can be used to irreversibly label dead cells prior to cryopreservation, fixation, and/or permeabilization procedures.

  • Unlike 7-AAD and propidium iodide, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized, and stained for intracellular antigens without any loss of staining intensity of the dead cells
  • Thus, using Fixable Viability Dyes allows dead cells to be excluded from analysis when intracellular targets are being studied
  • Fixable Viability Dye eFluor™ 780 can be excited by the red (633 nm) laser line and has a peak emission of 780 nm that can be detected using a 780/60 band pass filter (equivalent to APC-eFluor™ 780 or APC-Alexa Fluor™ 750)
  • For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye
  • If the percentage of dead cells is expected to be less than 5%, then it is recommended to take a small aliquot of cells and heat them at 65°C for 1 minute then immediately place on ice for 1 minute
  • After this treatment, the heat-killed cells can be combined 1:1 with live cells and then stained with the Fixable Viability Dye
  • Testing suggests that compensation out of most detectors is negligible, with compensation out of PE-Cyanine7 being the highest at less than 5%
  • Actual compensation values will depend on each investigator's specific instrument, filter sets, and PMT voltage settings
  • Fixable Viability Dye eFluor™ 780 is supplied as a pre-diluted solution prepared in high-quality, anhydrous DMSO
  • It should be protected from light and moisture
  • Store at -70°C with dessicant, it may be freeze-thawed up to 20 times
  • Allow vial to equilibrate to room temperature before opening
TRUSTED_SUSTAINABILITY

Specifications

Color Infrared
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Dye
Product Line eBioscience
Quantity 100 Tests
Emission Wavelength Range 633/780
How do I discriminate between live cells and dead cells when doing a flow cytometry analysis?

Discrimination between viable and non-viable cells can be carried out with the use of the 7-AAD Viability Dye (Cat. No. 00-6993-50) or Propidium Iodide (PI) Staining Solution (Cat. No. 00-6990-50). The 7-AAD or PI will mark the non-viable cells by binding to the nuclei of those cells. The nucleic acid of viable cells will not be accessible to the dye and will not be stained. When analyzing the data collected, gate out all cells stained with the viability dye. When staining for intracellular proteins, use the Fixable Viability Dye eFluor 455UV, 450, 520, 660, or 780.

Can Fixable Viability Dyes be used with the eBioscience BrdU Staining kits for Flow Cytometry?

Yes, a Fixable Viability Dye can be added to samples after BrdU labeling and before fixation with the BrdU Staining Buffer. See the protocol for additional information.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.